  • Search Results
  • The pluripotency factor nanog promotes breast cancer tumorigenesis and metastasis.
The pluripotency factor nanog promotes breast cancer tumorigenesis and metastasis.
Oncogene (2013-06-19)
X Lu, S J Mazur, T Lin, E Appella, Y Xu
ABSTRACT
Nanog is a transcription factor required for maintaining the pluripotency of embryonic stem cells, and is not expressed in most normal adult tissues. However, recent studies have indicated that Nanog is overexpressed in many types of human cancers, including breast cancer. To elucidate the physiological roles of Nanog in tumorigenesis, we developed an inducible Nanog transgenic mouse model, in which the expression of Nanog in adult tissues can be induced via LoxP/Cre-mediated deletion. Our findings indicate that overexpression of Nanog in the mammary gland is not sufficient to induce mammary tumor. However, when coexpressed with Wnt-1 in the mouse mammary gland, it promotes mammary tumorigenesis and metastasis. In this context, Nanog promotes the migration and invasion of breast cancer cells. Microarray analysis has shown that the ectopic expression of Nanog deregulates the expression of numerous genes associated with tumorigenesis and metastasis, such as the PDGFRα gene. Our findings demonstrate the involvement of Nanog in breast cancer metastasis, and provide the basis for the reported correlation between Nanog expression and poor prognosis of human breast cancer patients. As Nanog is not expressed in most adult tissues, these findings identify Nanog as a potential therapeutic target in the treatment of Nanog-expressing metastatic breast cancer.
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
o-Xylene, suitable for HPLC, 98%
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Methyl salicylate, United States Pharmacopeia (USP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)