  • Search Results
  • Transport of polymeric nanoparticulate drug delivery systems in the proximity of silica and sand.
Transport of polymeric nanoparticulate drug delivery systems in the proximity of silica and sand.
Environmental science & technology (2015-02-20)
I-Cheng Chen, Ming Zhang, Blake Teipel, Isa Silveira de Araujo, Yagmur Yegin, Mustafa Akbulut
ABSTRACT
The contamination of the environment with traditional therapeutics due to metabolic excretion, improper disposal, and industrial waste has been well-recognized. However, knowledge of the environmental distribution and fate of emerging classes of nanomedicine is scarce. This work investigates the effect of surface chemistry of polymeric nanoparticulate drug delivery systems (PNDDS) on their adsorption dynamics and transport in the vicinity of environmentally relevant surfaces for a concentration comparable with hospital and pharmaceutical manufacturing effluents. To this end, five different types of paclitaxel-based nanomedicine having different polymer stabilizers were employed. Their transport behavior was characterized via quartz crystal microbalance, sand column, spectrofluorometry, and dynamic light scattering techniques. PNDDS having positive zeta-potential displayed strong adsorption onto silica surfaces and no mobility in porous media of quartz sand, even in the presence of humic acid. The mobility of negatively charged PNDDS strongly depended on the amount and type of salt present in the aqueous media: Without any salt, such PNDDS demonstrated no adsorption on silica surfaces and high levels of mobility in sand columns. The presence of CaCl2 and CaSO4, even at low ionic strengths (i.e. 10 mM), induced PNDDS adsorption on silica surfaces and strongly limited the mobility of such PNDSS in sand columns.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%