  • Search Results
  • Successful nucleofection of rat adipose-derived stroma cells with Ambystoma mexicanum epidermal lipoxygenase (AmbLOXe).
Successful nucleofection of rat adipose-derived stroma cells with Ambystoma mexicanum epidermal lipoxygenase (AmbLOXe).
Stem cell research & therapy (2014-10-11)
Angela Fülbier, Reinhild Schnabel, Stefanie Michael, Peter M Vogt, Sarah Strauß, Kerstin Reimers, Christine Radtke
ABSTRACT
Adipose-derived stroma cells (ASCs) are attractive cells for cell-based gene therapy but are generally difficult to transfect. Nucleofection has proven to be an efficient method for transfection of primary cells. Therefore, we used this technique to transfect ASCs with a vector encoding for Ambystoma mexicanum epidermal lipoxygenase (AmbLOXe) which is a promising bioactive enzyme in regenerative processes. Thereby, we thought to even further increase the large regenerative potential of the ASCs. ASCs were isolated from the inguinal fat pad of Lewis rats and were subsequently transfected in passage 1 using Nucleofector® 2b and the hMSC Nucleofector kit. Transfection efficiency was determined measuring co-transfected green fluorescent protein (GFP) in a flow cytometer and gene expression in transfected cells was detected by reverse transcription polymerase chain reaction (RT-PCR). Moreover, cell migration was assessed using a scratch assay and results were tested for statistical significance with ANOVA followed by Bonferroni's post hoc test. High initial transfection rates were achieved with an average of 79.8 ± 2.82% of GFP positive cells although longer cultivation periods reduced the number of positive cells to below 5% after four passages. Although successful production of AmbLOXe transcript could be proven the gene product had no measureable effect on cell migration. Our study demonstrates the feasibility of ASCs to serve as a vehicle of AmbLOXe transport for gene therapeutic purposes in regenerative medicine. One potential field of applications could be peripheral nerve injuries.
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,4-Diaminobutane, 99%
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Supelco
Putrescine, analytical standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications