  • Search Results
  • Circulating TNF receptors 1 and 2 are associated with the severity of renal interstitial fibrosis in IgA nephropathy.
Circulating TNF receptors 1 and 2 are associated with the severity of renal interstitial fibrosis in IgA nephropathy.
PloS one (2015-04-11)
Yuji Sonoda, Tomohito Gohda, Yusuke Suzuki, Keisuke Omote, Masanori Ishizaka, Joe Matsuoka, Yasuhiko Tomino
ABSTRACT
The current study aimed to examine whether the levels of TNF receptors 1 and 2 (TNFR1 and TNFR2) in serum and urine were associated with other markers of kidney injury and renal histological findings, including TNFR expression, in IgA nephropathy (IgAN). The levels of the parameters of interest were measured by immunoassay in 106 biopsy-proven IgAN patients using samples obtained immediately before renal biopsy and in 34 healthy subjects. Renal histological findings were evaluated using immunohistochemistry. The levels of serum TNFRs were higher in IgAN patients than in healthy subjects. The levels of both TNFRs in serum or urine were strongly correlated with each other (r > 0.9). Serum TNFR levels were positively correlated with the urinary protein to creatinine ratio (UPCR) and four markers of tubular damage of interest (N-acetyl-β-D-glucosaminidase [NAG], β2 microglobulin [β2m], liver-type fatty acid-binding protein [L-FABP], and kidney injury molecule-1 [KIM-1]) and negatively correlated with estimated glomerular filtration rate (eGFR). Patients in the highest tertile of serum TNFR levels showed more severe renal interstitial fibrosis than did those in the lowest or second tertiles. The tubulointerstitial TNFR2-, but not TNFR1-, positive area was significantly correlated with the serum levels of TNFRs and eGFR. Stepwise multiple regression analysis revealed that elevated serum TNFR1 or TNFR2 levels were a significant determinant of renal interstitial fibrosis after adjusting for eGFR, UPCR, and other markers of tubular damage. In conclusion, elevated serum TNFR levels were significantly associated with the severity of renal interstitial fibrosis in IgAN patients. However, the source of TNFRs in serum and urine remains unclear.
Product Description
Supelco
Dibutyl phthalate, Selectophore
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)