  • Search Results
  • Alteration in cytochrome P450 3A4 activity as measured by a urine cortisol assay in HIV-1-infected pregnant women and relationship to antiretroviral pharmacokinetics.
Alteration in cytochrome P450 3A4 activity as measured by a urine cortisol assay in HIV-1-infected pregnant women and relationship to antiretroviral pharmacokinetics.
HIV medicine (2014-11-20)
F T Aweeka, C Hu, L Huang, B M Best, A Stek, P Lizak, S K Burchett, J S Read, H Watts, M Mirochnick, E V Capparelli
ABSTRACT
Pregnancy results in physiological changes altering the pharmacokinetics of drugs metabolized by cytochrome P450 3A4 (CYP3A4). The urinary ratio of 6-β hydroxycortisol to cortisol (6βHF : F) is a marker of CYP3A4 induction. We sought to evaluate its change in antiretroviral (ARV)-treated HIV-1-infected women and to relate this change to ARV pharmacokinetics. Women receiving various ARVs had pharmacokinetic evaluations during the third trimester of pregnancy (>30 weeks) and postpartum with determination of 6βHF : F carried out on the same days. The Wilcoxon signed rank test was used to compare the ratio antepartum to postpartum. The relationship between the change in ratio and the change in pharmacokinetics was analysed using Kendall's tau. 6βHF : F ratios were available for 107 women antepartum, with 54 having postpartum values. The ratio was higher antepartum (P=0.033) (median comparison 1.35; 95% confidence interval 1.01, 1.81). For 71 women taking a protease inhibitor (PI), the antepartum vs. postpartum 6βHF : F comparison was marginally significant (P=0.058). When the change in the 6βHF : F ratio was related to the change in the dose-adjusted ARV area under the plasma concentration vs. time curve (AUC) between antepartum and postpartum, the 35 subjects in the lopinavir/ritonavir (LPV/r) arms demonstrated an inverse relationship (P=0.125), albeit this correlation did not reach statistical significance. A 35% increase in the urinary 6βHF : F ratio was measured during late pregnancy compared with postpartum, indicating that CYP3A induction occurs during pregnancy. The trend towards an inverse relationship between the change in the 6βHF : F ratio and the change in the LPV AUC antepartum vs. postpartum suggests that CYP3A induction may be one mechanism behind altered LPV exposure during pregnancy.
Product Description
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard