Immune changes and dysphoric moods across the postpartum.
American journal of reproductive immunology (New York, N.Y. : 1989) (2014-09-18)
Maureen E Groer, Cecilia Jevitt, Ming Ji
ABSTRACT
Little is known about postpartum immune recovery and relationships of common dysphoric moods, stress, immunology, and endocrinology. Healthy women (n = 72) were followed for six postpartum months with immune and hormone measures and dysphoric moods and stress scales. A panel of cytokines produced in mitogen-stimulated whole blood assays were measured at each time, along with plasma levels of hsC-reactive protein (hsCRP), Interleukin-6 (IL-6), and a panel of hormones. Cellular immunity, measured by production of Interferon-gamma (IFNγ) and (Interleukin-2 (IL-2) from stimulated whole blood culture, was low in the early postpartum with changes by 3 months. Tumor necrosis factor alpha (TNFα) showed a similar pattern. Plasma levels of CRP and Interleukin-6 (IL-6) showed higher levels in the early postpartum. Mood disturbance scores dropped across the postpartum with a change in slope at 3 months. No significant relationships were found between immune, endocrine, and psychosocial measures. Return to normal cellular immune function may take 3-4 months in the postpartum. Some aspects of early immunology (hsCRP and IL-6) probably reflect the latter stage of pregnancy, the stress of birth and the inflammation associated with involution. Dysphoric moods are higher in the early postpartum but are not related to immune factors or hormones.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard