  • Search Results
  • Fascin is involved in the chemotherapeutic resistance of breast cancer cells predominantly via the PI3K/Akt pathway.
Fascin is involved in the chemotherapeutic resistance of breast cancer cells predominantly via the PI3K/Akt pathway.
British journal of cancer (2014-08-15)
H Ghebeh, S Al-Khaldi, S Olabi, A Al-Dhfyan, F Al-Mohanna, R Barnawi, A Tulbah, T Al-Tweigeri, D Ajarim, M Al-Alwan
ABSTRACT
A major therapeutic challenge for breast cancer is the ability of cancer cells to evade killing of conventional chemotherapeutic agents. We have recently reported the actin-bundling protein (fascin) as a major regulator of breast cancer metastasis and survival. Survival of breast cancer patients that received chemotherapy and xenograft tumour model was used to assess the effect of chemotherapy on fascin-positive and -negative breast cancer cells. Molecular and cellular assays were used to gain in-depth understanding of the relationship between fascin and chemoresistance. We showed a significant correlation between fascin expression and shorter survival in breast cancer patients who received chemotherapy. In xenograft experiments, fascin-positive cancer cells displayed significantly more resistance to chemotherapy-mediated apoptotic cell death than fascin-negative counterparts. This increased chemoresistance was at least partially mediated through PI3K/Akt signalling, and was paralleled by increased FAK phosphorylation, enhanced expression of the inhibitor of apoptosis proteins (XIAP and Livin) and suppression of the proapoptotic markers (caspase 9, caspase 3 and PARP). This is the first report to demonstrate fascin involvement in breast cancer chemotherapeutic resistance, supporting the development of fascin-targeting drugs for better treatment of chemoresistance breast cancer.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
Hematoxylin
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis