  • Search Results
  • Ndfip1 attenuated 6-OHDA-induced iron accumulation via regulating the degradation of DMT1.
Ndfip1 attenuated 6-OHDA-induced iron accumulation via regulating the degradation of DMT1.
Neurobiology of aging (2014-12-04)
Wenting Jia, Huamin Xu, Xixun Du, Hong Jiang, Junxia Xie
ABSTRACT
Elevated iron levels and increased expression of divalent metal transporter 1 (DMT1) in the substantia nigra of Parkinson's disease (PD) have been reported. Nedd4 family-interacting protein 1 (Ndfip1), an adaptor protein for the Nedd4 family of ubiquitin ligases, played an essential role in regulating DMT1 and iron homeostasis in human cortical neurons. In this study, we demonstrated that the expression of Ndfip1 decreased in 6-hydroxydopamine (6-OHDA)-induced PD rats and 6-OHDA-treated MES23.5 dopaminergic cells. Further study showed that the decrease of Ndfip1 occurred earlier than the increase of DMT1 with iron-responsive element (DMT1 + IRE) in 6-OHDA-treated MES23.5 cells, indicating that the decrease of Ndfip1 might be involved in the increase of DMT1 + IRE. In addition, we demonstrated that overexpression of Ndfip1 caused DMT1 + IRE downregulation, resulting in the decreased iron influx and iron-induced neurotoxicity. Although Ndfip1 knockdown led to decreased protein levels of DMT1 + IRE, partially aggravated iron-induced neurotoxicity. Further experiments showed that 6-OHDA-induced decrease in Ndfip1 levels might be related to proteasomal and lysosomal activations and oxidative stress caused by 6-OHDA. These data suggest that decreased Ndfip1 expression might contribute to the pathogenesis of 6-OHDA-induced iron accumulation and Ndfip1 could attenuate 6-OHDA-induced iron accumulation via regulating the degradation of DMT1.
Product Description
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)