Ca(2+) homeostasis in sealed t-tubules of mouse ventricular myocytes.
Journal of molecular and cellular cardiology (2014-05-03)
I Moench, A N Lopatin
ABSTRACT
We have recently shown that in mouse ventricular myocytes, t-tubules can be quickly and tightly sealed during the resolution of hyposmotic shock of physiologically relevant magnitude. Sealing of t-tubules is associated with trapping extracellular solution inside the myocytes but the ionic homeostasis of sealed t-tubules and the consequences of potential transtubular ion fluxes remain unknown. In this study we investigated the dynamics of Ca(2+) movements associated with sealing of t-tubules. The data show that under normal conditions sealed t-tubules contain Ca(2+) at concentrations below 100μM. However, blockade of voltage-dependent Ca(2+) channels with 10μM nicardipine, or increasing extracellular concentration of K(+) from 5.4mM to 20mM led to several fold increase in concentration of t-tubular Ca(2+). Alternatively, the release of Ca(2+) from sarcoplasmic reticulum using 10mM caffeine led to the restoration of t-tubular Ca(2+) towards extracellular levels within few seconds. Sealing of t-tubules in the presence of extracellular 1.5mM Ca(2+) and 5.4mM extracellular K(+) led to occasional and sporadic intracellular Ca(2+) transients. In contrast, sealing of t-tubules in the presence of 10mM caffeine was characterized by a significant long lasting increase in intracellular Ca(2+). The effect was completely abolished in the absence of extracellular Ca(2+) and significantly reduced in pre-detubulated myocytes but was essentially preserved in the presence of mitochondrial decoupler dinitrophenol. This study shows that sealed t-tubules are capable of highly regulated transport of Ca(2+) and present a major route for Ca(2+) influx into the cytosol during sealing process.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
2-Methyl-2-butanol, ReagentPlus®, 99%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Caffeine, certified reference material, TraceCERT®
Sigma-Aldrich
2-Methyl-2-butanol, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methyl-2-butanol, anhydrous, ≥99%
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Methyl-2-butanol, analytical standard