  • Search Results
  • Effects of neonatal corticosterone and environmental enrichment on retinal ERK1/2 and CREB phosphorylation in adult mice.
Effects of neonatal corticosterone and environmental enrichment on retinal ERK1/2 and CREB phosphorylation in adult mice.
Experimental eye research (2014-08-26)
Andrea Matteucci, Chiara Ceci, Cinzia Mallozzi, Simone Macrì, Fiorella Malchiodi-Albedi, Giovanni Laviola
ABSTRACT
Exposure to Stimulating Environments (SE) during development may improve neuroplasticity in central nervous system, protect against neurotoxic damage, and promote neuronal recovery in adult life. While biochemical mechanisms of SE-promoted neuronal plasticity are well known in the brain, much less is known on the signaling cascade governing plasticity and neuroprotection in the retina. In order to investigate if in the retina signaling molecules involved in neuronal plasticity are affected by SE, neonatal CD-1 mice were exposed to moderate corticosterone levels (NC), supplemented through maternal milk during the first postnatal week, or to environmental enrichment (EE) conditions (physical and social stimuli) from early adolescence. Our results showed that both NC and EE increased the phosphorylation level of Extracellularly Regulated Kinase 1/2 (ERK1/2) and cAMP response element-binding protein (CREB) in the adult retinal tissue. Furthermore, we observed that activated ERK1/2 was restricted to Müller cells, while pCREB was mostly present in the nuclei of retinal neurons. Neither NC, nor EE modified the expression of GFAP, a marker of Müller cells activation. In conclusion our results indicate that both NC and EE activate ERK1/2 and CREB in the retina and provide a biochemical background for the neuroprotective activity exerted by SE against retinal damage. Furthermore, they support the role of Müller glia as a key cell determinant of retinal neuroplasticity.
Sigma-Aldrich
Anti-GFAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GFAP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GFAP antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MNK1 (pThr255) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GFAP antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GFAP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-MAPK3 (pThr202) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MAPK3 (pTyr204) antibody produced in rabbit, affinity isolated antibody