  • Search Results
  • Comprehensive bioimaging with fluorinated nanoparticles using breathable liquids.
Comprehensive bioimaging with fluorinated nanoparticles using breathable liquids.
Nature communications (2015-01-21)
Michael E Kurczy, Zheng-Jiang Zhu, Julijana Ivanisevic, Adam M Schuyler, Kush Lalwani, Antonio F Santidrian, John M David, Anand Giddabasappa, Amanda J Roberts, Hernando J Olivos, Peter J O'Brien, Lauren Franco, Matthew W Fields, Liliana P Paris, Martin Friedlander, Caroline H Johnson, Adrian A Epstein, Howard E Gendelman, Malcolm R Wood, Brunhilde H Felding, Gary J Patti, Mary E Spilker, Gary Siuzdak
ABSTRACT
Fluorocarbons are lipophobic and non-polar molecules that exhibit remarkable biocompatibility, with applications in liquid ventilation and synthetic blood. The unique properties of these compounds have also enabled mass spectrometry imaging of tissues where the fluorocarbons act as a Teflon-like coating for nanostructured surfaces to assist in desorption/ionization. Here we report fluorinated gold nanoparticles (f-AuNPs) designed to facilitate nanostructure imaging mass spectrometry. Irradiation of f-AuNPs results in the release of the fluorocarbon ligands providing a driving force for analyte desorption. The f-AuNPs allow for the mass spectrometry analysis of both lipophilic and polar (central carbon) metabolites. An important property of AuNPs is that they also act as contrast agents for X-ray microtomography and electron microscopy, a feature we have exploited by infusing f-AuNPs into tissue via fluorocarbon liquids to facilitate multimodal (molecular and anatomical) imaging.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Toluene, analytical standard
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Perfluorodecalin, 95%
Sigma-Aldrich
1-Octanethiol, ≥98.5%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetradecafluorohexane, 99%
Sigma-Aldrich
1H,1H,2H,2H-Perfluorodecanethiol, 97%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanethiol, ≥96.0%
Sigma-Aldrich
Octadecafluorodecahydronaphthalene, (cis+trans), ≥94% (GC)
Sigma-Aldrich
Tetradecafluorohexane, A mixture of perfluorinated hexanes, 95%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Toluene, HPLC grade, 99.8%