Hinokitiol is a novel glycoprotein VI antagonist on human platelets.
Platelets (2014-01-18)
Wan-Jung Lu, Ming-Ping Wu, Kuan-Hung Lin, Yu-Chen Lin, Hsiu-Chu Chou, Joen-Rong Sheu
ABSTRACT
Hinokitiol (4-isopropyl-tropolone) is a bioactive compound with various pharmacological activities that is found in the wood of cupressaceous plants. Platelet activation plays an important role in thrombogenesis. In our previous study, hinokitiol specifically inhibited collagen-induced platelet aggregation ex vivo and prolonged thrombogenesis in vivo. The glycoprotein (GP) VI and integrin α2β1 are major collagen receptors that mediate platelet adhesion and aggregation. In our current study, we investigated which of these collagen receptors is involved in the hinokitiol-mediated inhibition of platelet activation. Treatment with 2-100 µM hinokitiol caused a dose-dependent right, parallel shift in the collagen concentration-response curve (0.5-10 µg/ml), with no change in the maximal responses. Furthermore, hinokitiol inhibited platelet aggregation and relative [Ca(2+)]i mobilization stimulated by convulxin, an agonist of GP VI, but not by aggretin, an agonist of integrin α2β1, indicating that hinokitiol mediates the inhibition of platelet activation through GP VI, rather than through integrin α2β1. Hinokitiol also specifically inhibited the convulxin-mediated activation of protein kinase C, phospholipase Cγ2, Akt, mitogen-activated protein kinases, and Lyn. Hinokitiol markedly diminished the co-immunoprecipitation of GP VI-bound Lyn after convulxin stimulation. In conclusion, hinokitiol, an antagonist of collagen GP VI may represent a novel antiplatelet drug for the prevention of thrombi associated with coronary and cerebral artery diseases.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Fura 2-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture