  • Search Results
  • Lizard tail regeneration: regulation of two distinct cartilage regions by Indian hedgehog.
Lizard tail regeneration: regulation of two distinct cartilage regions by Indian hedgehog.
Developmental biology (2015-01-18)
Thomas P Lozito, Rocky S Tuan
ABSTRACT
Lizards capable of caudal autotomy exhibit the remarkable ability to "drop" and then regenerate their tails. However, the regenerated lizard tail (RLT) is known as an "imperfect replicate" due to several key anatomical differences compared to the original tail. Most striking of these "imperfections" concerns the skeleton; instead of the vertebrae of the original tail, the skeleton of the RLT takes the form of an unsegmented cartilage tube (CT). Here we have performed the first detailed staging of skeletal development of the RLT CT, identifying two distinct mineralization events. CTs isolated from RLTs of various ages were analyzed by micro-computed tomography to characterize mineralization, and to correlate skeletal development with expression of endochondral ossification markers evaluated by histology and immunohistochemistry. During early tail regeneration, shortly after CT formation, the extreme proximal CT in direct contact with the most terminal vertebra of the original tail develops a growth plate-like region that undergoes endochondral ossification. Proximal CT chondrocytes enlarge, express hypertrophic markers, including Indian hedgehog (Ihh), apoptose, and are replaced by bone. During later stages of tail regeneration, the distal CT mineralizes without endochondral ossification. The sub-perichondrium of the distal CT expresses Ihh, and the perichondrium directly calcifies without cartilage growth plate formation. The calcified CT perichondrium also contains a population of stem/progenitor cells that forms new cartilage in response to TGF-β stimulation. Treatment with the Ihh inhibitor cyclopamine inhibited both proximal CT ossification and distal CT calcification. Thus, while the two mineralization events are spatially, temporally, and mechanistically very different, they both involve Ihh. Taken together, these results suggest that Ihh regulates CT mineralization during two distinct stages of lizard tail regeneration.
Product Description
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Supelco
Hematoxylin
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)