  • Search Results
  • Alteration in the expression of exon IIC transcripts of brain-derived neurotrophic factor gene by simvastatin [correction of simvastain] in chronic mild stress in mice: a possible link with dopaminergic pathway.
Alteration in the expression of exon IIC transcripts of brain-derived neurotrophic factor gene by simvastatin [correction of simvastain] in chronic mild stress in mice: a possible link with dopaminergic pathway.
Canadian journal of physiology and pharmacology (2014-11-13)
Digvijay G Rana, Amrutlal K Patel, Chaitanya G Joshi, Mayurdhvaj K Jhala, Ramesh K Goyal
ABSTRACT
We have investigated the influence of dopaminergic agents on the expression of brain-derived neurotrophic factor (BDNF) gene in relation with lipid levels in chronic mild stress (CMS). Mice subjected to CMS were treated with simvastatin (10 mg/kg, per os (orally)) along with bromocriptine (2 mg/kg, intraperitoneally (ip)), levodopa (200 mg/kg, ip), or haloperidol (0.1 mg/kg, ip) for 14 days. CMS produced a decrease in sucrose intake and an increase in serum cholesterol and triglycerides levels with a decrease in high-density lipoprotein cholesterol, which were prevented by simvastatin. This was greater when it was combined with bromocriptine or levodopa. Haloperidol significantly prevented the simvastatin-induced increase in sucrose intake but not the alterations in lipids. There was an upregulation in the expression of BDNF exon-IIA and -IIB transcripts by CMS but not the exon-IIC transcripts. Simvastatin could increase expression of exon-IIC transcripts in stressed mice. This was partially increased by bromocriptine. Haloperidol significantly prevented simvastatin-induced increase in expression of BDNF exon-IIC transcripts. The results showed a positive correlation between expression of BDNF exon-IIC transcripts and sucrose intake. In conclusion, our data suggest the involvement of lipid levels and BDNF exon-IIC transcripts in CMS-induced behaviour in mice, possibly through the dopaminergic system.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Haloperidol, powder
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
DL-3-Hydroxy-3-methylglutaryl coenzyme A sodium salt hydrate, ≥90% (HPLC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Simvastatin, analytical standard
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Levodopa, European Pharmacopoeia (EP) Reference Standard
Haloperidol, European Pharmacopoeia (EP) Reference Standard
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
Simvastatin, European Pharmacopoeia (EP) Reference Standard