  • Search Results
  • The phosphodiesterase 3 inhibitor cilostazol does not stimulate growth of colorectal liver metastases after major hepatectomy.
The phosphodiesterase 3 inhibitor cilostazol does not stimulate growth of colorectal liver metastases after major hepatectomy.
Clinical & experimental metastasis (2014-07-24)
Moritz J Strowitzki, Stefan Dold, Maximilian von Heesen, Christina Körbel, Claudia Scheuer, Mohammed R Moussavian, Martin K Schilling, Otto Kollmar, Michael D Menger
ABSTRACT
Liver failure after extended hepatectomy represents a major challenge in the surgery of hepatic colorectal metastasis. A previous study has indicated that inhibition of phosphodiesterase type 3 (PDE 3) stimulates liver regeneration. However, little is known whether PDE 3 inhibitors, such as cilostazol, also stimulate the growth of remnant metastases. Therefore, we herein studied the effect of cilostazol on engraftment, vascularization and growth of colorectal liver metastasis after major hepatectomy. WAG-rats underwent either major hepatectomy or sham operation. Metastases were induced by subcapsular implantation of 5 × 10(5) CC531-colorectal cancer cells. Animals were daily treated with cilostazol (5 mg/kg body weight) or glucose solution. Tumor growth was measured by high-resolution ultrasound at days 7 and 14. Tumor vascularization and tumor cell proliferation were determined by immunohistochemistry and western blotting. High-resolution ultrasound analysis in hepatectomized and non-hepatectomized animals showed that cilostazol does not stimulate tumor growth. Accordingly, the number of PCNA-positive tumor cells did not differ between cilostazol-treated animals and sham-treated controls. Interestingly, cilostazol reduced tumor vascularization in both hepatectomized and non-hepatectomized animals. This was indicated by a significantly lower number of platelet-endothelial cell adhesion molecule (PECAM-1)-positive cells in tumors of cilostazol-treated animals compared to sham-treated controls. The PDE 3 inhibitor cilostazol does not stimulate the growth of colorectal metastases during liver regeneration after major hepatectomy.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri