  • Search Results
  • Water stress proteins from Nostoc commune Vauch. exhibit anti-colon cancer activities in vitro and in vivo.
Water stress proteins from Nostoc commune Vauch. exhibit anti-colon cancer activities in vitro and in vivo.
Journal of agricultural and food chemistry (2014-12-20)
Songjia Guo, Shuhua Shan, Xiaoting Jin, Zongwei Li, Zhuoyu Li, Liangqi Zhao, Quan An, Wei Zhang
ABSTRACT
Nostoc commune has been traditionally used in China as a health food and medicine. The water stress proteins (WSP) of Nostoc commune are the major component of the extracellular matrix. This study purified and identified the water stress proteins (WSP1) from Nostoc commune Vauch., which could inhibit the proliferation of human colon cancer cell lines. The IC50 values of WSP1 against DLD1, HCT116, HT29, and SW480 cells were 0.19 ± 0.02, 0.21 ± 0.03, 0.39 ± 0.05, and 0.41 ± 0.01 μg/μL, respectively. Notably, it displayed very little effect on the normal human intestinal epithelial FHC cell line. The IC50 value of WSP1 against FHC cells was 0.67 ± 0.05 μg/μL. Moreover, the growth of DLD1 xenografted tumors in nude mice were significantly suppressed in the WSP1 treated group. Mechanistically, the cell-cycle analysis revealed that WSP1 induced growth inhibition by G1/S arrest. Meanwhile, Western blotting and immunohistochemistry assays showed WSP1 could activate caspase-8, -9, and -3, along with subsequent PARP cleavage. Furthermore, the pan-caspase inhibitor, z-VAD-FMK, partly reversed the effect caused by WSP1, confirming that WSP1 induced cell apoptosis through caspase-dependent pathway. Collectively, WSP1 has targeted inhibition for colon cancer proliferation both in vitro and in vivo and it is valuable for future exploitation and utilization as an antitumor agent.
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O