  • Search Results
  • In vitro/in vivo screening of oxidative homeostasis and damage to DNA, protein, and lipids using UPLC/MS-MS.
In vitro/in vivo screening of oxidative homeostasis and damage to DNA, protein, and lipids using UPLC/MS-MS.
Analytical and bioanalytical chemistry (2014-06-28)
Aitor Carretero, Zacarías León, Juan Carlos García-Cañaveras, Angela Zaragoza, María José Gómez-Lechón, María Teresa Donato, Agustín Lahoz
ABSTRACT
Multiple analytical methods are required to comprehensively assess oxidative homeostasis and specific damage to macromolecules. Our aim was to develop a straightforward strategy for the fast assessment of global oxidative status and specific damage to DNA, proteins, and lipids. To this end, an analytical method, based on ultra-performance liquid chromatography coupled to mass spectrometry (UPLC-MS/MS), was developed and validated for the quantification of 16 oxidative stress (OS) biomarkers. Some of these markers were unstable; thus, an easy sample treatment procedure, including fractionation and derivatization, was set up. The method was validated according to Food and Drug Administration (FDA) guidelines, and it provided good results in terms of intra- and inter-day precision (≤17.2 and 16 %, respectively), accuracy (relative error measurement between -16.6 and 19.8 %), and linearity (R (2) > 0.994). The approach was applied to determine the oxidative insult provoked to cultured rat hepatocytes by cumene hydroperoxide and to analyze the liver and serum samples from patients diagnosed with nonalcoholic steatohepatitis. In both studies, significant differences were found if compared to the corresponding control groups; interestingly, ophthalmic acid was shown as an OS biomarker in both models for the first time. A key advantage of the novel approach in comparison with former multi-method approaches is that now a single method is applied to assess the 16 OS biomarkers. Its comprehensive capacity to profile oxidative homeostasis and damage in both in vitro and clinical samples has been illustrated, which indicates that the proposed approach is a good choice to evaluate whether OS is involved in physiological signals, diseases, or toxic events and to what extent.
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets