  • Search Results
  • Comparative evaluation of N-acetylcysteine and N-acetylcysteineamide in acetaminophen-induced hepatotoxicity in human hepatoma HepaRG cells.
Comparative evaluation of N-acetylcysteine and N-acetylcysteineamide in acetaminophen-induced hepatotoxicity in human hepatoma HepaRG cells.
Experimental biology and medicine (Maywood, N.J.) (2014-09-24)
Shakila Tobwala, Ahdab Khayyat, Weili Fan, Nuran Ercal
ABSTRACT
Acetaminophen (N-acetyl-p-aminophenol, APAP) is one of the most widely used over-the-counter antipyretic analgesic medications. Despite being safe at therapeutic doses, an accidental or intentional overdose can result in severe hepatotoxicity; a leading cause of drug-induced liver failure in the U.S. Depletion of glutathione (GSH) is implicated as an initiating event in APAP-induced toxicity. N-acetylcysteine (NAC), a GSH precursor, is the only currently approved antidote for an APAP overdose. Unfortunately, fairly high doses and longer treatment times are required due to its poor bioavailability. In addition, oral and intravenous administration of NAC in a hospital setting are laborious and costly. Therefore, we studied the protective effects of N-acetylcysteineamide (NACA), a novel antioxidant, with higher bioavailability and compared it with NAC in APAP-induced hepatotoxicity in a human-relevant in vitro system, HepaRG. Our results indicated that exposure of HepaRG cells to APAP resulted in GSH depletion, reactive oxygen species (ROS) formation, increased lipid peroxidation, mitochondrial dysfunction (assessed by JC-1 fluorescence), and lactate dehydrogenase release. Both NAC and NACA protected against APAP-induced hepatotoxicity by restoring GSH levels, scavenging ROS, inhibiting lipid peroxidation, and preserving mitochondrial membrane potential. However, NACA was better than NAC at combating oxidative stress and protecting against APAP-induced damage. The higher efficiency of NACA in protecting cells against APAP-induced toxicity suggests that NACA can be developed into a promising therapeutic option for treatment of an APAP overdose.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized disodium salt, suitable for cell culture, BioReagent
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard