  • Search Results
  • Do venepuncture procedures induce cortisol responses? A review, study, and synthesis for stress research.
Do venepuncture procedures induce cortisol responses? A review, study, and synthesis for stress research.
Psychoneuroendocrinology (2014-06-03)
Lisa J Weckesser, Franziska Plessow, Maximilian Pilhatsch, Markus Muehlhan, Clemens Kirschbaum, Robert Miller
ABSTRACT
Venepuncture procedures are frequently employed to continuously monitor humoral stress markers. As such procedures are conceived as "potent psychological and physiological stressors", there is a need to determine whether venepuncture procedures themselves elicit cortisol responses and if so, how to deal with them appropriately. In order to assess the rate of cortisol responses to venepuncture, we conducted a literature review, which suggested that venepuncture procedures induce cortisol responses with a probability of approximately 30%. By utilizing Bayesian analysis, this result was integrated with the cortisol data of 18 healthy men who were exposed to a venepuncture procedure twice (time lag: 1 week). The currently observed response rate of 47% differed substantially from the earlier findings, which we attribute to a self-selective sampling of participants. In addition, participants showing a response to the first venepuncture were highly likely to also show a response to the second one. In this regard, we discuss the presumed conditioning of cortisol responses to venepuncture procedures. To prevent the superposition of venepuncture-induced cortisol responses and responses induced by target stressors, we propose a time- and selection-based strategy: cortisol samples taken about 110min after venepuncture should be virtually adjusted for its superimposing effects. Furthermore, previous experiences of venepuncture were highly predictive for cortisol responsiveness. This association could be utilized in further studies to identify participants who will probably show a cortisol response to venepuncture.
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard