  • Search Results
  • Vitrification of a small number of spermatozoa in normozoospermic and severely oligozoospermic samples.
Vitrification of a small number of spermatozoa in normozoospermic and severely oligozoospermic samples.
Systems biology in reproductive medicine (2014-11-22)
Valeriy Kuznyetsov, Sergey I Moskovtsev, Michael Crowe, Ayub G-M Lulat, Clifford L Librach
ABSTRACT
Despite broad utilization of sperm cryopreservation, little progress has been made to modify freezing protocols or to improve rates of sperm survival. Vitrification is an alternative method for freezing human spermatozoa without toxic permeable cryoprotectants (CPAs). The purpose of our study was to optimize the vitrification and post thaw recovery of a small number of spermatozoa using only nonpermeating CPAs in a closed straw system in normozoospermic and severely oligozoospermic samples. Individual motile spermatozoa (n = 295) were selected from semen samples of 15 normozoospermic and 10 severe oligozoospermia patients. Overall sperm recovery after vitrification was 80% (n = 236) with 80% (n = 189) viability and 41.5% (n = 98) retained post-warming motility. Two different loading techniques were compared to transfer selected spermatozoa into straws in preparation for vitrification: by spontaneous capillary action (CA) and with the aid of a polar body biopsy (PBB) pipette. There was evidence that the PBB loading technique increases the odds of spermatozoa recovery in both subsets (p = 0.01 and p = 0.04) in the normal and abnormal subsamples, respectively.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20