  • Search Results
  • Interaction of arginine with protein during refolding process probed by amide H/D exchange mass spectrometry and isothermal titration calorimetry.
Interaction of arginine with protein during refolding process probed by amide H/D exchange mass spectrometry and isothermal titration calorimetry.
Biochimica et biophysica acta (2014-12-03)
Dawei Zhao, Yongdong Liu, Guifeng Zhang, Chun Zhang, Xiunan Li, Qingqing Wang, Hong Shi, Zhiguo Su
ABSTRACT
Arginine has been widely used as low molecular weight additive to promote protein refolding by suppressing aggregate formation. However, methods to investigate the role of arginine in protein refolding are often limited on protein's global conformational properties. Here, hydrogen/deuterium exchange mass spectrometry (HDX-MS) was used to study the effects of arginine on recombinant human granulocyte colony-stimulating factor (rhG-CSF) refolding at the scale of peptide mapping. It was found that deuteration levels of rhG-CSF refolded with arginine was higher than that without arginine during the whole refolding process, but they became almost the same when the refolding reached equilibrium. This phenomenon indicated that arginine could protect some amide deuterium atoms from being exchanged with hydrogen, but the protection diminished gradually along with refolding proceeding. Enzymatic digestion revealed six particular peptides of 16-47, 72-84, 84-93, 114-124, 145-153 and 154-162 were mainly responsible for the deuteration, and all of them dominantly located in protein's α-helix domain. Furthermore, thermodynamics analysis by isothermal titration calorimetry provided direct evidence that arginine could only react with denatured and partially refolded rhG-CSF. Taking all of the results together, we suggest that arginine suppresses protein aggregation by a reversible combination. At the initial refolding stage, arginine could combine with the denatured protein mainly through hydrogen bonding. Subsequently, arginine is gradually excluded from protein with protein's native conformation recovering.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)