  • Search Results
  • Formulation, stability and pharmacokinetics of sugar-based salmon calcitonin-loaded nanoporous/nanoparticulate microparticles (NPMPs) for inhalation.
Formulation, stability and pharmacokinetics of sugar-based salmon calcitonin-loaded nanoporous/nanoparticulate microparticles (NPMPs) for inhalation.
International journal of pharmaceutics (2015-02-11)
Maria Inês Amaro, Frederic Tewes, Oliviero Gobbo, Lidia Tajber, Owen I Corrigan, Carsten Ehrhardt, Anne Marie Healy
ABSTRACT
A challenge exists to produce dry powder inhaler (DPI) formulations with appropriate formulation stability, biological activity and suitable physicochemical and aerosolisation characteristics that provide a viable alternative to parenteral formulations. The present study aimed to produce sugar-based nanoporous/nanoparticulate microparticles (NPMPs) loaded with a therapeutic peptide - salmon calcitonin (sCT). The physicochemical properties of the powders and their suitability for pulmonary delivery of sCT were determined. Production of powders composed of sCT loaded into raffinose or trehalose with or without hydroxypropyl-β-cyclodextrin was carried out using a laboratory scale spray dryer. Spray dried microparticles were spherical, porous and of small geometric size (≤2 μm). Aerodynamic assessment showed that the fine particle fraction (FPF) less than 5 μm ranged from 45 to 86%, depending on the formulation. The mass median aerodynamic diameter (MMAD) varied between 1.9 and 4.7 μm. Compared to unprocessed sCT, sCT:raffinose composite systems presented a bioactivity of approximately 100% and sCT:trehalose composite systems between 70-90% after spray drying. Storage stability studies demonstrated composite systems with raffinose to be more stable than those containing trehalose. These sugar-based salmon calcitonin-loaded NPMPs retain reasonable sCT bioactivity and have micromeritic and physicochemical properties which indicate their suitability for pulmonary delivery. Formulations presented a similar pharmacokinetic profile to sCT solution. Hence the advantage of a dry powder formulation is its non-invasive delivery route and ease of administration of the sCT.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%