  • Search Results
  • MHC class II presentation is controlled by the lysosomal small GTPase, Arl8b.
MHC class II presentation is controlled by the lysosomal small GTPase, Arl8b.
Journal of immunology (Baltimore, Md. : 1950) (2015-02-01)
Xavier Michelet, Salil Garg, Benjamin J Wolf, Amit Tuli, Paola Ricciardi-Castagnoli, Michael B Brenner
ABSTRACT
Dendritic cells (DCs) are specialized APCs with the ability to prime naive T cells. DCs first sample Ags from the environment and then orchestrate their processing and loading onto MHC class II (MHC II) Ag-presenting molecules in lysosomes. Once MHC II molecules have bound a peptide, the MHC II-peptide complex is delivered to the cell surface for presentation to CD4(+) T cells. Regulation of Ag uptake via macropinocytosis and phagocytosis has been extensively studied, as well as trafficking in early endocytic vesicles notably regulated by the small GTPase Rab5 and its effectors. However, little is known about the regulators of Ag delivery from early endosomes to lysosomal compartments where the proper pH, proteases, MHC II, invariant chain, and HLA-DM reside, awaiting exogenous Ags for loading. In this article, we report the crucial role of the small GTPase ADP-ribosylation factor-like 8b (Arl8b) in MHC II presentation in DCs. We show for the first time, to our knowledge, that Arl8b localizes to MHC II compartments in DCs and regulates formation of MHC II-peptide complexes. Arl8b-silenced DCs display a defect in MHC II-Ag complex formation and its delivery to the cell surface during infection resulting in a defect in T cell recognition. Our results highlight the role of Arl8b as a trafficking regulator of the late stage of complex formation and MHC II presentation in DCs.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
HEPES