  • Search Results
  • Digestive development in neonatal dairy calves with either whole or ground oats in the calf starter.
Digestive development in neonatal dairy calves with either whole or ground oats in the calf starter.
Journal of dairy science (2015-03-10)
F X Suarez-Mena, A J Heinrichs, C M Jones, T M Hill, J D Quigley
ABSTRACT
A series of 3 trials was conducted to determine effects of whole or ground oats in starter grain on reticulorumen fermentation and digestive system development of preweaned calves. Male Holstein calves (43.1±2.3kg at birth; n=8, 9, and 7 for trials 1, 2, and 3, respectively) were housed in individual pens in a heated facility; bedding was covered with landscape fabric to prevent consumption of bedding by the calves. In trials 1 and 2 only, calves were fitted with rumen cannulas by wk 2 of life. In all trials, a fixed amount of starter (containing 25% oats either ground and in the pellet or whole) was offered daily; orts were fed through the cannula in trials 1 and 2. Calves were randomly assigned to an all-pelleted starter or pellets plus whole oats. Rumen contents (trials 1 and 2) were sampled weekly at -8, -4, 0, 2, 4, 8, and 12 h after grain feeding for determination of pH and volatile fatty acids. Calves were killed 3 wk (trial 1) or 4 wk (trials 2 and 3) after grain was offered; organs were harvested, emptied, rinsed, and weighed to gauge digestive organ development. Starter intake was not different between treatments. Weekly measurements of rumen digesta pH did not change and only subtle changes were observed in molar proportions of individual volatile fatty acids. Molar proportion of butyrate and pH linearly decreased with age, whereas acetate proportion increased. Reticulorumen weight and papillae length tended to be greater for calves fed pelleted starter, whereas abomasum weight was greater for calves fed pellets plus whole oats. Fecal particle size and starch content were greater for calves fed pellets plus whole oats. Under the conditions of this study, physical form of oats in starter grain did not affect rumen fermentation measurements; greater rumen weight and papillae length in calves fed pelleted starter may be the result of greater nutrient availability of ground oats. Under the conditions of this study with young calves on treatments for <4 wk, increasing particle size of the starter by feeding whole oats did not affect rumen fermentation nor did it improve digestive system development.
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Pyridoxine, ≥98%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Calcium phosphate dibasic, 98.0-105.0%
Supelco
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Folic acid, meets USP testing specifications
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%