  • Search Results
  • DNA-PK/Chk2 induces centrosome amplification during prolonged replication stress.
DNA-PK/Chk2 induces centrosome amplification during prolonged replication stress.
Oncogene (2014-03-26)
C-Y Wang, E Y-H Huang, S-C Huang, B-C Chung
ABSTRACT
The antineoplastic drug hydroxyurea (HU), when used at subtoxic doses, induces prolonged replication stress and centrosome amplification. This causes genomic instability and increases the malignancy of the recurring tumor. The mechanism of centrosome amplification induced by prolonged replication stress, however, is still unclear. Here, we examined the involvement of ataxia telangiectasia, mutated (ATM), ataxia telangiectasia, mutated and Rad3-related (ATR) and DNA-dependent protein kinase (DNA-PK) and found that HU-induced centrosome amplification was inhibited by the depletion of DNA-PKcs, but not ATM and ATR. Inactivation of ATM/ATR in U2OS cells instead caused aneuploidy and cell death. We found DNA-PKcs depletion also abrogated ATM phosphorylation, indicating that ATM activation during prolonged replication stress depends on DNA-PK. Depletion of DNA-PK abrogated checkpoint kinase (Chk)2 activation and partially reduced Chk1 activation. Chk2 depletion blocked HU-induced centrosome amplification, indicating a function of Chk2 in centrosome amplification. We further found that Chk2 was phosphorylated at Thr68 on the mother centriole at late G2 and mitosis when unstressed and on all amplified centrioles induced by HU. In summary, we have elucidated that DNA-PK/Chk2 signaling induces centrosome amplification upon long-term HU treatment, therefore increasing our insight into tumor recurrence after initial chemotherapy.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)