  • Search Results
  • Synthesis, characterization and in vitro and in vivo investigation of C₃F₈-filled poly(lactic-co-glycolic acid) nanoparticles as an ultrasound contrast agent.
Synthesis, characterization and in vitro and in vivo investigation of C₃F₈-filled poly(lactic-co-glycolic acid) nanoparticles as an ultrasound contrast agent.
Molecular medicine reports (2014-11-15)
Cui-Wei Wang, Shi-Ping Yang, He Hu, Jing Du, Feng-Hua Li
ABSTRACT
The present study aimed to prepare perfluoropropane (C3F8)-filled poly(lactic-co-glycolic acid) (PLGA) nanoparticles and investigate the feasibility of using PLGA nanoparticles as an ultrasound contrast agent. The PLGA nanoscale ultrasound contrast agent was prepared using a modified double-emulsion solvent evaporation method. Camphor in the form of a sublimable porogen was added to render the nanoparticles hollow and enable C3F8 gas introduction. Various physicochemical properties of PLGA nanoparticles, including morphology, size and dispersion, were analyzed by electron microscopy and dynamic laser scattering. In vitro ultrasound imaging of C3F8-filled PLGA nanoparticles was also investigated under various imaging conditions. Further in vivo ultrasound imaging was conducted on male rats following intratesticular injection of PLGA nanoparticles. C3F8-filled PLGA nanoparticles with a mean diameter of 152.0±58.08 nm were obtained. Electron microscopy revealed spherical-shaped nanoparticles with smooth surfaces, a capsular morphology and a large hollow within. In vitro ultrasound imaging of hollow PLGA nanoparticles indicated marked signal enhancement. Local intensity of the acoustical signal continued to increase during PLGA-nanoparticle injection into the testicle and the ability of hollow PLGA nanoparticles to enhance ultrasound imaging in vivo was demonstrated. The enhancement image of testicular tissue following injection with C3F8-filled PLGA nanoparticles was sustained for a minimum of five minutes. In conclusion, the hollow C3F8-filled PLGA nanoparticles were demonstrated to have potential for applications as a novel ultrasound contrast agent.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Supelco
2-Propanol, analytical standard
Supelco
Dichloromethane, analytical standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer