  • Search Results
  • Understanding red blood cell parameters in the context of the frailty phenotype: interpretations of the FIBRA (Frailty in Brazilian Seniors) study.
Understanding red blood cell parameters in the context of the frailty phenotype: interpretations of the FIBRA (Frailty in Brazilian Seniors) study.
Archives of gerontology and geriatrics (2014-09-23)
João Carlos Silva, Zélia Vieira de Moraes, Conceição Silva, Silvia de Barros Mazon, Maria Elena Guariento, Anita Liberalesso Neri, André Fattori
ABSTRACT
Frailty and anemia in the elderly appear to share a common pathophysiology associated with chronic inflammatory processes. This study uses an analytical, cross-sectional, population-based methodology to investigate the probable relationships between frailty, red blood cell parameters and inflammatory markers in 255 community-dwelling elders aged 65 years or older. The frailty phenotype was assessed by non-intentional weight loss, fatigue, low grip strength, low energy expenditure and reduced gait speed. Blood sample analyses were performed to determine hemoglobin level, hematocrit and reticulocyte count, as well as the inflammatory variables IL-6, IL-1ra and hsCRP. In the first multivariate analysis (model I), considering only the erythroid parameters, Hb concentration was a significant variable for both general frailty status and weight loss: a 1.0g/dL drop in serum Hb concentration represented a 2.02-fold increase (CI 1.12-3.63) in an individual's chance of being frail. In the second analysis (model II), which also included inflammatory cytokine levels, hsCRP was independently selected as a significant variable. Each additional year of age represented a 1.21-fold increase in the chance of being frail, and each 1-unit increase in serum hsCRP represented a 3.64-fold increase in the chance of having the frailty phenotype. In model II reticulocyte counts were associated with weight loss and reduced metabolic expenditure criteria. Our findings suggest that reduced Hb concentration, reduced RetAbs count and elevated serum hsCRP levels should be considered components of frailty, which in turn is correlated with sarcopenia, as evidenced by weight loss.
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant