  • Search Results
  • Semi-continuous, label-free immunosensing approach for Ca2+-based conformation change of a calcium-binding protein.
Semi-continuous, label-free immunosensing approach for Ca2+-based conformation change of a calcium-binding protein.
The Analyst (2014-06-10)
Sung-Ho Paek, Ji-Na Park, Dong-Hyung Kim, Hee-Soo Kim, Un-Hwan Ha, Sung-Kyu Seo, Se-Hwan Paek
ABSTRACT
A label-free immunosensing method based on the conformational change of calcium-binding protein (CBP) depending on analyte concentration was explored for semi-continuous analysis of free Ca(2+). Glucose-galactose-binding protein as a CBP and produced as a recombinant protein by Escherichia coli was used as the immunogen to produce monoclonal antibodies by hybridoma technology. We finally screened the 3-6F cell clone, which produced the desired antibody specific to a particular structural conformation of the protein that occurred only upon CBP-calcium complex formation. To construct an immunosensor, the antibody was immobilized via a secondary antibody on an Octet Red optical fiber-based label-free sensor. Calcium analysis was conducted on the sensor in combination with CBP previously added to the aqueous sample, which distinguished the sensor signal according to the analyte concentration. The immunosensor produced a signal in real time with a response time of approximately 15 min and could be reused for analyses of different samples in a semi-continuous manner. The minimum detection limit of the analyte under optimal conditions was 0.09 mM and the upper limit was about 5 mM (log-logit transformed standard curve linearity: R(2) > 98%). In sample tests with milk, the analytical performance of the sensor was highly correlated (R(2) > 99%) with that of the reference system based on the KMnO4 titration method (ISO 12081). Although the sensor showed cross-reactivity at high concentrations (>1 mM) of cations including zinc, iron, manganese, and copper, these ionic components were not traceable (<0.01 mM) in milk.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Aminopterin, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Aminopterin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Aminopterin, powder
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)