  • Search Results
  • Effect of light irradiation by light emitting diode on colon cancer cells.
Effect of light irradiation by light emitting diode on colon cancer cells.
Anticancer research (2014-09-10)
Noriko Matsumoto, Kozo Yoshikawa, Mitsuo Shimada, Nobuhiro Kurita, Hirohiko Sato, Takashi Iwata, Jun Higashijima, Motoya Chikakiyo, Masaaki Nishi, Hideya Kashihara, Chie Takasu, Shohei Eto, Akira Takahashi, Masatake Akutagawa, Takahiro Emoto
ABSTRACT
Recent studies have demonstrated the efficacy of irradiation from light emitting diodes (LED) for wound healing, anti-inflammation and anticancer therapies. However, little is known about the effects of visible light in colon cancer cells. The purpose of this study was to evaluate the biological response (including gene expression changes) of human colon cancer cells to different wavelengths of LED irradiation. Human colon cancer cells (HT29 or HCT116) were seeded onto laboratory dishes that were then put on LED irradiation equipment with a 465 nm-, 525 nm-, or 635 nm-LED. Irradiation at 15 or 30 mW was performed 10 min/day, each day for 5 days. The cell counting kit8 was then used to measure cell viability. Apoptosis and expression of several mRNAs (caspase, MAPK and autophagy pathway) in HT29 cultures irradiated with 465 nm LED were evaluated via AnnexinV/PI and RT-PCR, respectively. Viability of HT29 and HCT116 cells was lower in 465 nm-LED irradiated cultures than in control cultures, but viability of HT29 cells did not differ between control cultures and 525 nm-LED or 635 nm-LED irradiated cultures. Moreover, the expression of FAS, caspase-3, capase-8, and JUK were significantly higher in 465 nm-LED irradiated cultures than in control cultures, and expression of ERK1/2 and LC3 was lower in blue-irradiated cells. LED irradiation at 465 nm inhibited the proliferation of HT29 cells and of HCT116 cells. Notably, LED irradiation at 465 nm promoted apoptosis inHT29 cultures via the extrinsic apoptosis pathway and the MAPK pathway.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)