  • Search Results
  • p53 and mitogen-activated protein kinase pathway protein profiles in fresh and frozen spermatozoa.
p53 and mitogen-activated protein kinase pathway protein profiles in fresh and frozen spermatozoa.
Andrologia (2013-12-10)
S Karabulut, A Demiroğlu-Zergeroğlu, E Yılmaz, F Sağır, N Delikara
ABSTRACT
Sperm or testicular tissue cryopreservation is performed in cases of male infertility as a treatment for the preservation of fertility. When these sperm cells are used in assisted reproductive techniques, fertilisation rates, developmental and implantation potential of embryos decrease and the abortion rates increase. In the present work, differences of both phosphorylation and expression levels of p53 and Mitogen-activated protein kinases (MAPK) proteins were analysed in 61 individual sperm samples before and after cryopreservation. We observed that p53 protein residue at Ser 15 was phosphorylated after cryopreservation. Because MAPK pathway activations may be involved in p53 phosphorylation, MAPK/ERK, Stress-activated protein kinases (SAPK)/JNK and p38MAPK proteins were also investigated. Analysis showed that p38MAPK phosphorylations increased significantly. However, ERK and JNK expressions and phosphorylations decreased, although the differences were not statistically significant. According to our results, it may be suggested that cryopreservation process activates p53 via p38 MAPK pathway that subsequently causes apoptosis, which may be related to sperm parameters.
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Synaptophysin antibody, Mouse monoclonal, clone SVP-38, purified from hybridoma cell culture
Sigma-Aldrich
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-TRP53 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MNK1 (pThr255) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TP53 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MAPK8 antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-MAPK8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-JNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-MAPK3 (pThr202) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MAPK3 (pTyr204) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody