  • Search Results
  • Correlation between glucose uptake and membrane potential in Leishmania parasites isolated from DCL patients with therapeutic failure: a proof of concept.
Correlation between glucose uptake and membrane potential in Leishmania parasites isolated from DCL patients with therapeutic failure: a proof of concept.
Parasitology research (2014-03-29)
Maritza Padrón-Nieves, Claudia Machuca, Emilia Díaz, Paulo Cotrim, Noris Rodriguez, Alicia Ponte-Sucre
ABSTRACT
Besides infection with drug-resistant parasites, therapeutic failure in leishmaniasis may be caused by altered drug pharmacokinetics, re-infection, and host immunologic compromise. Our aim has been to evaluate if relapses that occur in patients suffering from diffuse cutaneous leishmaniasis (DCL) associate with changes in the fitness of infecting organisms. Therefore, in isolates from patients suffering DCL, we correlated glucose uptake and plasma membrane potential and compared the results with those obtained from reference strains. The data demonstrate that Leishmania parasites causing DCL incorporate glucose at an efficient rate, albeit without significant changes in the plasma membrane potential as their corresponding reference strains. The isolate that did not change its accumulation rate of glucose compared to its reference strain expressed a less polarized membrane potential that was insensitive to mitochondrial inhibitors, suggesting a metabolic dysfunction that may result in glycolysis being the main source of ATP. The results constitute a proof of concept that indicates that parasites causing DCL adapted well to drug pressure and expressed an increased fitness. That is, that in Leishmania mexicana and Leishmania amazonensis, parasites isolated from DCL patients, a strong modification of the parasite physiology might occur. As consequences, the parasites adapted well to drug pressure, increased their fitness, and they had an efficient glucose uptake rate albeit not significant changes in membrane potential as their corresponding reference strains. Further validation of the concepts herein established and whether or not the third isolate corresponds with a drug-resistant phenotype need to be demonstrated at the genetic level.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%