  • Search Results
  • TLR4-dependent activation of dendritic cells by an HMGB1-derived peptide adjuvant.
TLR4-dependent activation of dendritic cells by an HMGB1-derived peptide adjuvant.
Journal of translational medicine (2014-08-16)
Rebecca Saenz, Diahnn Futalan, Lien Leutenez, Fien Eekhout, Jessie F Fecteau, Simeon Sundelius, Stig Sundqvist, Marie Larsson, Tomoko Hayashi, Boris Minev, Dennis Carson, Sadik Esener, Bradley Messmer, Davorka Messmer
ABSTRACT
High mobility group box protein 1 (HMGB1) acts as an endogenous danger molecule that is released from necrotic cells and activated macrophages. We have previously shown that peptide Hp91, whose sequence corresponds to an area within the B-Box domain of HMGB1, activates dendritic cells (DCs) and acts as an adjuvant in vivo. Here we investigated the underlying mechanisms of Hp91-mediated DC activation. Hp91-induced secretion of IL-6 was dependent on clathrin- and dynamin-driven endocytosis of Hp91 and mediated through a MyD88- and TLR4-dependent pathway involving p38 MAPK and NFκB. Endosomal TLR4 has been shown to activate the MyD88-independent interferon pathway. Hp91-induced activation of pIRF3 and IL-6 secretion was reduced in IFNαβR knockout DCs, suggesting an amplification loop via the IFNαβR. These findings elucidate the mechanisms by which Hp91 acts as immunostimulatory peptide and may serve as a guide for the future development of synthetic Th1-type peptide adjuvants for vaccines.
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
SAFC
BIS-TRIS
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
SAFC
HEPES
SAFC
L-Glutamine
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-p-Tosyl-L-phenylalanine chloromethyl ketone, ≥97% (TLC), powder