  • Search Results
  • Antiproliferative and quinone reductase-inducing activities of withanolides derivatives.
Antiproliferative and quinone reductase-inducing activities of withanolides derivatives.
European journal of medicinal chemistry (2014-06-01)
Manuela E García, Viviana E Nicotra, Juan C Oberti, Carla Ríos-Luci, Leticia G León, Laura Marler, Guannan Li, John M Pezzuto, Richard B van Breemen, José M Padrón, Idaira Hueso-Falcón, Ana Estévez-Braun
ABSTRACT
Two new and five known withanolides (jaborosalactones 2, 3, 4, 5, and 24) were isolated from the leaves of Jaborosa runcinata Lam. We also obtained some derivatives from jaborosalactone 5, which resulted to be the major isolated metabolite. The natural compounds as well as derivatives were evaluated for their antiproliferative activity and the induction of quinone reductase 1 (QR1; NQ01) activity. Structure-activity relationships revealed valuable information on the pharmacophore of withanolide-type compounds. Three compounds of this series showed significantly higher antiproliferative activity than jaborosalactone 5. The effect of these compounds on the cell cycle was determined. Furthermore, the ability of major compounds to induce QR1 was evaluated. It was found that all the active test compounds are monofunctional inducers that interact with Keap1. The most promising derivatives prepared from jaborosalactone 5 include (23R)-4β,12β,21-trihydroxy-1,22-dioxo-12,23-cycloergostan-2,5,17,24-tetraen-26,23-olide (18) and (23R)-21-acetoxy-12β-hydroxy-1,22-dioxo-12,23-cycloergostan-2,5,17,24-tetraen-26,23-lactame (20).
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
Dimethyl sulfoxide, analytical standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O