  • Search Results
  • Protective effect of Ferula gummosa hydroalcoholic extract against nitric oxide deficiency-induced oxidative stress and inflammation in rats renal tissues.
Protective effect of Ferula gummosa hydroalcoholic extract against nitric oxide deficiency-induced oxidative stress and inflammation in rats renal tissues.
Clinical and experimental hypertension (New York, N.Y. : 1993) (2014-05-03)
Seyed Jafar Moosavi, Masoumeh Habibian, Maghsoud Peeri, Mohammad Ali Azarbayjani, Seyed Mohammad Nabavi, Seyed Fazel Nabavi, Antoni Sureda
ABSTRACT
Nitric oxide (NO) synthase inhibition increases hypertension and causes renal injury. Ferula gummosa is used in Iranian traditional medicine for treatment of several diseases and has been reported to exert a potent anti-inflammatory and antioxidant action. The aim of this investigation was to evaluate the renoprotective effects of hydroalcoholic extract of Ferula gummosa (HEG) on Nω-nitro-L-arginine methyl ester (L-NAME)-induced oxidative stress and inflammation and explore the mechanisms that link NO deficiency with altered renal heat shock protein (HSP70). Rats were injected intraperitoneally with L-NAME (10 mg/kg) to induce renal injury. Simultaneously, HEG (90 mg/kg) was administered by gastric gavage to L-NAME-treated rats for 6 days/week during an 8-week period. Renal thiobarbituric acid reactive substances (TBARS), superoxide dismutase (SOD), interleukin-6 (IL-6), tumor necrosis factor alpha (TNF-α), HSP70, plasma NO and total antioxidant capacity (TAC) were evaluated. The administration of L-NAME significantly increased renal TBARS, TNF-α, IL-6, HSP70 levels and decreased renal SOD activity, that these changes were accompanied by the reduced plasma NO and TAC levels. HEG administration decreased TBARS, HSP70, TNF-α and IL-6 levels and increased SOD activity in the kidney tissues of L-NAME treated rats (p<0.05). Also, plasma TAC level and NO bioavailability have been elevated after administration of HEG (p<0.05). These findings support that NO deficiency induces renal stress oxidative and inflammation, which markedly increased renal HSP70 and HEG could protect kidney against these damaging effects via its anti-oxidative, anti-inflammatory action and modulate renal HSP70.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Pefabloc® SC, analytical standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard