  • Search Results
  • EGCG inhibits properties of glioma stem-like cells and synergizes with temozolomide through downregulation of P-glycoprotein inhibition.
EGCG inhibits properties of glioma stem-like cells and synergizes with temozolomide through downregulation of P-glycoprotein inhibition.
Journal of neuro-oncology (2014-09-01)
Yong Zhang, Shao-Xiang Wang, Ji-Wei Ma, Hai-Ying Li, Jie-Cheng Ye, Si-Ming Xie, Bin Du, Xue-Yun Zhong
ABSTRACT
Combination therapy to inhibit cancer stem cells may have important clinical implications. Here, we examine the molecular mechanisms by which epigallocatechin gallate (EGCG), a bioactive polyphenol in green tea, inhibits the stem cell characteristics of glioma stem-like cells (GSLCs) and synergizes with temozolomide (TMZ), a DNA-methylating agent commonly used as first-line chemotherapy in gliomas. GSLCs were enriched from the human glioblastoma cell line U87 using neurosphere culture. Cells were analyzed using flow cytometry, quantitative PCR, and western blotting. Compared to U87 cells, a higher percentage of U87 GSLCs remained in the G0/G1 phase, with downregulation of the cell-cycle protein CylinD1 and overexpression of stem cell markers CD133 and ALDH1. The drug-resistance gene ABCB1 (but not ABCG2 or MGMT) also showed high mRNA and protein expression. The resistance index of U87 GSLCs against TMZ and carmustine (BCNU) was 3.0 and 16.8, respectively. These results indicate that U87 GSLCs possess neural stem cell and drug-resistance properties. Interestingly, EGCG treatment inhibited cell viability, neurosphere formation, and migration in this cell model. EGCG also induced apoptosis, downregulation of p-Akt and Bcl-2, and cleaving PARP in a dose-dependent manner. Importantly, EGCG treatment significantly downregulated P-glycoprotein expression but not that of ABCG2 or MGMT and simultaneously enhanced sensitivity to TMZ. Our study demonstrates that the use of EGCG alone or in combination with TMZ may be an effective therapeutic strategy for glioma.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard
Epigallocatechin gallate, primary reference standard