  • Search Results
  • Neonatal thymectomy favors Helicobacter pylori-promoted gastric mucosa-associated lymphoid tissue lymphoma lesions in BALB/c mice.
Neonatal thymectomy favors Helicobacter pylori-promoted gastric mucosa-associated lymphoid tissue lymphoma lesions in BALB/c mice.
The American journal of pathology (2014-06-10)
Delphine Chrisment, Pierre Dubus, Lucie Chambonnier, Anaïs Hocès de la Guardia, Elodie Sifré, Alban Giese, Myriam Capone, Camille Khairallah, Pierre Costet, Benoît Rousseau, Christophe Hubert, Odile Burlen-Defranoux, Christine Varon, Antonio Bandeira, Francis Mégraud, Philippe Lehours
ABSTRACT
Neonatal thymectomy in BALB/c mice has been described as a model of gastric mucosa-associated lymphoid tissue (MALT) lymphoma (GML). By using this experimental system, we screened, for the first time to our knowledge, Helicobacter pylori GML-associated strains for their capacity to promote disease. A cohort of BALB/c mice underwent thymectomy at day 3 after birth (d3Tx). Successful thymic ablation was evaluated by the degree of lymphopenia in blood samples collected at 4 weeks of age. d3Tx and non-thymectomized controls were infected with either GML strains (B38 or B47) or control strains (SS1 or TN2GF4). Gastric samples collected at 6, 12, and 18 months after infection were studied for bacteria content, and submitted to histological, immunochemical, molecular, and immunological analyses. Severe gastric inflammation was only observed in d3Tx mice. In these animals, the gastric lamina propria was infiltrated with lymphoid cells organized in follicles composed of B cells with few infiltrating T cells. PCR of D/J IgH gene segments proved the monoclonality of infiltrating B cells, which strongly correlated with the presence of lymphoepithelial lesions. B-cell infiltrates were particularly prominent in mice infected with the B47-GML strain. No pathological changes were detected in noninfected d3Tx mice. We identified new H. pylori isolates adapted to the mouse stomach with high potential of GML development, which is only revealed in hosts rendered lymphopenic by neonatal thymic ablation.
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, for molecular biology, ≥99.0%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
1,4-Dithioerythritol, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Monoclonal Anti-CD45 antibody produced in rat, clone EM-05, purified immunoglobulin, buffered aqueous solution
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications