  • Search Results
  • Active immunisation of mice with GnRH lipopeptide vaccine candidates: Importance of T helper or multi-dimer GnRH epitope.
Active immunisation of mice with GnRH lipopeptide vaccine candidates: Importance of T helper or multi-dimer GnRH epitope.
Bioorganic & medicinal chemistry (2014-07-26)
Daryn Goodwin, Pavla Simerska, Cheng-Hung Chang, Friederike M Mansfeld, Pegah Varamini, Michael J D'Occhio, Istvan Toth
ABSTRACT
Active immunisation against gonadotropin releasing hormone (GnRH) is a potential alternative to surgical castration. This study focused on the development of a GnRH subunit lipopeptide vaccine. A library of vaccine candidates that contained one or more (up to eight) copies of monomeric or dimeric GnRH peptide antigen, an adjuvanting lipidic moiety based on lipoamino acids, and an additional T helper epitope, was synthesised by solid phase peptide synthesis. The candidates were evaluated in vivo in order to determine the minimal components of this vaccine necessary to induce a systemic immune response. BALB/c mice were immunised with GnRH lipopeptide conjugates, co-administered with or without Complete Freund's Adjuvant, followed by two additional immunisations. Significant GnRH-specific IgG titres were detected in sera obtained from mice immunised with four of the seven lipopeptides tested, with an increase in titres observed after successive immunisations. This study highlights the importance of for epitope optimisation and delivery system design when producing anti-hapten antibodies in vivo. The results of this study also contribute to the development of future clinical and veterinary immunocontraceptives.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Human IgG ELISA Kit, for serum, plasma