  • Search Results
  • Intravenous GLP-1 (7-36) amide for prevention of hyperglycemia during cardiac surgery: a randomized, double-blind, placebo-controlled study.
Intravenous GLP-1 (7-36) amide for prevention of hyperglycemia during cardiac surgery: a randomized, double-blind, placebo-controlled study.
Journal of cardiothoracic and vascular anesthesia (2013-10-23)
Benjamin A Kohl, Mary S Hammond, Andrew J Cucchiara, E Andrew Ochroch
ABSTRACT
The authors sought to evaluate the efficacy of an intravenous glucagon-like peptide-1 (GLP-1) infusion, compared with placebo, to mitigate intraoperative hyperglycemia. Prospective, double-blinded, randomized, placebo-controlled. University hospital. Diabetic (non-insulin dependent) and non-diabetic patients undergoing elective cardiac surgery with cardiopulmonary bypass. Patients were randomized in a 1:1 fashion to GLP-1 (7-36) amide infusion (1.5 pmol/kg/min) or placebo. Insulin was administered intraoperatively to both groups per a standardized protocol. A total of 77 patients were included for analysis (GLP-1, n = 37; placebo, n = 40). Mean blood glucose during cardiopulmonary bypass was 127.5 mg/dL and 142.5 mg/dL (p = 0.002) in the GLP-1 and placebo groups, respectively. Mean blood glucose values during the entire intraoperative course were 12.2 mg/dL lower for subjects given GLP-1 (95% CI 2.3, 22, p = 0.015), independent of time. During the period of cardiopulmonary bypass, mean blood glucose values in subjects given GLP-1 were 14.1 mg/dL lower than those who received placebo (95% CI 3.5, 24.8, p = 0.009), independent of time. The incidence of hypoglycemia did not differ significantly between the 2 groups. Administration of intravenous GLP-1 (7-36) amide to patients undergoing cardiac surgery significantly reduced their plasma glucose levels intraoperatively and may represent a novel therapeutic strategy to prevent perioperative hyperglycemia.
Product Description
Serine, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard