  • Search Results
  • Non-specific SIRT inhibition as a mechanism for the cytotoxicity of ginkgolic acids and urushiols.
Non-specific SIRT inhibition as a mechanism for the cytotoxicity of ginkgolic acids and urushiols.
Toxicology letters (2014-07-08)
Lucie Ryckewaert, Lionel Sacconnay, Pierre-Alain Carrupt, Alessandra Nurisso, Claudia Simões-Pires
ABSTRACT
Ginkgolic acids and urushiols are natural alkylphenols known for their mutagenic, carcinogenic and genotoxic potential. However, the mechanism of toxicity of these compounds has not been thoroughly elucidated so far. Considering that the SIRT inhibitory potential of anacardic acids has been hypothesized by in silico techniques, we herein demonstrated through both in vitro and computational methods that structurally related compounds such as ginkgolic acids and urushiols are able to modulate SIRT activity. Moreover, their SIRT inhibitory profile and cytotoxicity were comparable to sirtinol, a non-specific SIRT inhibitor (SIRT1 and SIRT2), and different from EX-527, a SIRT1 specific inhibitor. This is the first report on the SIRT inhibition of ginkgolic acids and urushiols. The results reported here are in line with previously observed effects on the induction of apoptosis by this class of compounds, and the non-specific SIRT inhibition is suggested as a new mechanism for their in vitro cytotoxicity.
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sirtuin 1 human, recombinant, expressed in E. coli, ≥60% (SDS-PAGE)
Sigma-Aldrich
SIRT1 human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
Sirtuin 2 human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M