  • Search Results
  • Pre-clinical evaluation of AZD-2014, a novel mTORC1/2 dual inhibitor, against renal cell carcinoma.
Pre-clinical evaluation of AZD-2014, a novel mTORC1/2 dual inhibitor, against renal cell carcinoma.
Cancer letters (2014-12-03)
Bing Zheng, Jia-Hui Mao, Lin Qian, Hua Zhu, Dong-hua Gu, Xiao-dong Pan, Fang Yi, Dong-mei Ji
ABSTRACT
Here we found that dual mTORC1/2 inhibitor AZD-2014 significantly inhibited RCC cell survival and growth, with higher efficiency than conventional mTORC1 inhibitors rapamycin and RAD001. RCC cell apoptosis was also induced by AZD-2014. AZD-2014 disrupted mTORC1/2 assembly and activation, while downregulating HIF-1α/2α and cyclin D1 expressions in RCC cells. Meanwhile, AZD-2014 activated autophagy, detected by p62 degradation, Beclin-1/ATG-5 upregulation and light LC3B-I/-II conversion. Autophagy inhibition by pharmacologic or siRNA-based means increased AZD-2014 activity in vitro, causing substantial RCC cell apoptosis. In vivo, AZD-2014 was more efficient than RAD001 in inhibiting 786-0 xenografts and downregulating HIF-1α/2α or p-AKT (Ser-473). Finally, AZD-2014's activity in vivo was further enhanced by co-administration of the autophagy inhibitor 3-methyaldenine. We provide evidence for clinical trials of using AZD-2014 in RCC treatment.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
SAFC
HEPES
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)