  • Search Results
  • Nanosized silver (II) pyridoxine complex to cause greater inflammatory response and less cytotoxicity to RAW264.7 macrophage cells.
Nanosized silver (II) pyridoxine complex to cause greater inflammatory response and less cytotoxicity to RAW264.7 macrophage cells.
Nanoscale research letters (2015-04-09)
Avijit Paul, Hee Ju, Sabarinathan Rangasamy, Yumi Shim, Joon Myong Song
ABSTRACT
With advancements in nanotechnology, silver has been engineered into a nanometre size and has attracted great research interest for use in the treatment of wounds. Silver nanoparticles (AgNPs) have emerged as a potential alternative to conventional antibiotics because of their potential antimicrobial property. However, AgNPs also induce cytotoxicity, generate reactive oxygen species (ROS), and cause mitochondrial damage to human cells. Pyridoxine possesses antioxidant and cell proliferation activity. Therefore, in the present investigation, a nanosilver-pyridoxine complex (AgPyNP) was synthesized, and its cytotoxicity and immune response was compared with AgNPs in macrophage RAW264.7 cells. Results revealed that AgPyNPs showed less cytotoxicity compared with AgNPs by producing a smaller amount of ROS in RAW264.7 cells. Surprisingly, however, AgPyNPs caused macrophage RAW264.7 cells to secrete a larger amount of interleukin-8 (IL-8) and generate a more active inflammatory response compared to AgNPs. It activated TNF-α, NF-κB p65, and NF-κB p50 to generate a more vigorous immune protection that produces a greater amount of IL-8 compared to AgNPs. Overall findings indicate that AgPyNPs exhibited less cytotoxicity and evoked a greater immune response in macrophage RAW264.7 cells. Thus, it can be used as a better wound-healing agent than AgNPs. Graphical AbstractFigurative representation of the comparison of AgNPs and AgPyNPs in macrophage RAW264.7 cells in terms of cytotoxicity and immune response.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Dioctyl sulfosuccinate sodium salt, ≥97%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture