  • Search Results
  • Identification of apolipoprotein C-I as a potential Wilms' tumor marker after excluding inflammatory factors.
Identification of apolipoprotein C-I as a potential Wilms' tumor marker after excluding inflammatory factors.
International journal of molecular sciences (2014-09-16)
Junjie Zhang, Fei Guo, Lei Wang, Wei Zhao, Da Zhang, Heying Yang, Jiekai Yu, Lili Niu, Fuquan Yang, Shu Zheng, Jiaxiang Wang
ABSTRACT
Wilms' tumor is one of the most common malignant tumors observed in children, and its early diagnosis is important for late-stage treatment and prognosis. We previously screened and identified protein markers for Wilms' tumor; however, these markers lacked specificity, and some were associated with inflammation. In the current study, serum samples from children with Wilms' tumors were compared with those of healthy controls and patients with systemic inflammatory response syndrome (SIRS). After exclusion of factors associated with inflammation, specific protein markers for Wilms' tumors were identified. After comparing the protein peak values obtained from all three groups, a protein with a m/z of 6438 Da was specified. Purification and identification of the target protein using high-pressure liquid chromatography (HPLC) and two-dimensional liquid chromatography-linearion trap mass spectrometry(2D-LC-LTQ-MS) mass spectrometry, respectively, revealed that it was apolipoprotein C-I (APO C-I). Thus, APO C-I is a specific protein marker for Wilms' tumor.
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%