Natural melanin: a potential pH-responsive drug release device.
International journal of pharmaceutics (2014-04-29)
Marco Araújo, Raquel Viveiros, Tiago R Correia, Ilídio J Correia, Vasco D B Bonifácio, Teresa Casimiro, Ana Aguiar-Ricardo
ABSTRACT
This work proposes melanin as a new nanocarrier for pH-responsive drug release. Melanin is an abundant natural polymer that can be easily extracted from cuttlefish as nanoparticles with a suitable size range for drug delivery. However, despite its high potentiality, the application of this biopolymer in the pharmaceutical and biomedical fields is yet to be explored. Herein, melanin nanoparticles were impregnated with metronidazole, chosen as model antibiotic drug, using supercritical carbon dioxide. The drug release profile was investigated at acidic and physiologic pH, and the dominant mechanism was found to follow a non-Fickian transport. Drug release from melanin shows a strong pH dependency, which allied to its biocompatibility and lack of cytotoxicity envisages its potential application as nanocarrier in formulations for colon and intestine targeted drug delivery.
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Resazurin sodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine
Metronidazole, European Pharmacopoeia (EP) Reference Standard