  • Search Results
  • A novel strategy for the rapid preparation and isolation of intact immune complexes from peptide mixtures.
A novel strategy for the rapid preparation and isolation of intact immune complexes from peptide mixtures.
Journal of molecular recognition : JMR (2014-07-22)
Mahmoud Al-Majdoub, Kwabena F M Opuni, Yelena Yefremova, Cornelia Koy, Peter Lorenz, Reham F El-Kased, Hans-Jürgen Thiesen, Michael O Glocker
ABSTRACT
The development and application of a miniaturized affinity system for the preparation and release of intact immune complexes are demonstrated. Antibodies were reversibly affinity-adsorbed on pipette tips containing protein G´ and protein A, respectively. Antigen proteins were digested with proteases and peptide mixtures were exposed to attached antibodies; forming antibody-epitope complexes, that is, immune complexes. Elution with millimolar indole propionic acid (IPA)-containing buffers under neutral pH conditions allowed to effectively isolate the intact immune complexes in purified form. Size exclusion chromatography was performed to determine the integrity of the antibody-epitope complexes. Mass spectrometric analysis identified the epitope peptides in the respective SEC fractions. His-tag-containing recombinant human glucose-6-phosphate isomerase in combination with an anti-His-tag monoclonal antibody was instrumental to develop the method. Application was extended to the isolation of the intact antibody-epitope complex of a recombinant human tripartite motif 21 (rhTRIM21) auto-antigen in combination with a rabbit polyclonal anti-TRIM21 antibody. Peptide chip analysis showed that antibody-epitope binding of rhTRIM21 peptide antibody complexes was not affected by the presence of IPA in the elution buffer. By contrast, protein G´ showed an ion charge structure by electrospray mass spectrometry that resembled a denatured conformation when exposed to IPA-containing buffers. The advantages of this novel isolation strategy are low sample consumption and short experimental duration in addition to the direct and robust methodology that provides easy access to intact antibody-antigen complexes under neutral pH and low salt conditions for subsequent investigations.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
SAFC
BIS-TRIS
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlus®, 99%
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)
SAFC
BIS-TRIS
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)