  • Search Results
  • Prognostic value of aldosterone and cortisol in patients hospitalized for acutely decompensated chronic heart failure with and without mineralocorticoid receptor antagonism.
Prognostic value of aldosterone and cortisol in patients hospitalized for acutely decompensated chronic heart failure with and without mineralocorticoid receptor antagonism.
Journal of cardiac failure (2015-01-13)
Gülmisal Güder, Fabian Hammer, Timo Deutschbein, Susanne Brenner, Dominik Berliner, NiKolas Deubner, Martin Bidlingmaier, Georg Ertl, Bruno Allolio, Christiane E Angermann, Martin Fassnacht, Stefan Störk
ABSTRACT
Serum aldosterone and cortisol independently predict an increased mortality risk in heart failure (HF), and mineralocorticoid receptor antagonism (MRA) improves survival. The prognostic relevance of aldosterone and cortisol with MRA is unclear. In this post hoc analysis of a prospective cohort, study serum levels of aldosterone and cortisol were measured at baseline in 842 patients with systolic HF. The mean age was 68 ± 12 years (27% female, 45% in New York Heart Association functional class III/IV, 43% with MRA; median follow-up 38 months [interquartile range 30-43 mo]). Crude mortality in the total cohort was 43% (patients with vs without MRA: 34% vs 41%; P = .052). In MRA-naïve patients, higher levels of both aldosterone and cortisol were predictive of increased mortality risk in multivariable Cox regression: hazard ratio (HR) with 95% confidence interval of highest vs lowest tertile for aldosterone: 1.51 [1.02-2.24] (P = .040); and for cortisol: 1.94 [1.28-2.93] (P = .002). In MRA-treated patients, aldosterone (highest vs lowest tertile: HR 1.65 [1.01-2.71]; P = .048) but not cortisol (HR 0.77 [0.44-1.27]; P = .33) was associated with all-cause mortality. Further subgroup analysis revealed that particularly patients with low cortisol and high aldosterone levels had the worst prognosis (HR 5.01 [2.22-11.3]; P < .001), compared with the reference of low cortisol and low aldosterone. Subjects with this profile had larger ventricles and more often coronary artery disease. In systolic HF, the prognostic value of aldosterone and cortisol levels differs in dependency of MRA intake. The pathophysiologic link between low cortisol, high aldosterone, and increased mortality risk in patients with MRA needs to be clarified.
Product Description
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard