  • Search Results
  • Novel 4-thiazolidinones as non-nucleoside inhibitors of hepatitis C virus NS5B RNA-dependent RNA polymerase.
Novel 4-thiazolidinones as non-nucleoside inhibitors of hepatitis C virus NS5B RNA-dependent RNA polymerase.
Archiv der Pharmazie (2014-12-03)
Gizem Çakir, İlkay Küçükgüzel, Rupa Guhamazumder, Esra Tatar, Dinesh Manvar, Amartya Basu, Bhargav A Patel, Javairia Zia, Tanaji T Talele, Neerja Kaushik-Basu
ABSTRACT
In continuation of our efforts to develop new derivatives as hepatitis C virus (HCV) NS5B inhibitors, we synthesized novel 5-arylidene-4-thiazolidinones. The novel compounds 29-42, together with their synthetic precursors 22-28, were tested for HCV NS5B inhibitory activity; 12 of these compounds displayed IC50 values between 25.3 and 54.1 µM. Compound 33, an arylidene derivative, was found to be the most active compound in this series with an IC50 value of 25.3 µM. Molecular docking studies were performed on the thumb pocket-II of NS5B to postulate the binding mode for these compounds.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%