MilliporeSigma
ERBB3 as an independent prognostic marker for nasopharyngeal carcinoma.
Journal of clinical pathology (2014-05-16)
Warut Tulalamba, Noppadol Larbcharoensub, Tavan Janvilisri
ABSTRACT
Although the ERBB proteins have been shown to be associated in many types of human tumours and serve as important cancer therapeutic targets, however, data regarding the expression and clinical relevance of ERBBs in nasopharyngeal carcinoma (NPC) are still conflicting. The aim of this study is to investigate the expression pattern of all ERBB members simultaneously in NPC tissues using immunohistochemistry and determine their clinical relevance. The expression of all members of ERBB proteins was evaluated using immunohistochemistry on 82 NPC tissue samples. Relationship between the ERBB protein expression, clinicopathological parameters and patient outcome was assessed using univariate and multivariate analyses. We found that ERBB1, ERBB2 and ERBB3 were strongly expressed in the normal nasopharyngeal epithelial cells. A marked reduction of ERBB1 and ERBB2 expression in NPC was observed compared with the non-cancerous tissues. 76 of 82 (92.7%) cases were ERBB3-positive, while ERBB4 was not expressed in both normal and NPC. The univariate log-rank analysis showed that regional lymph node metastasis, systemic metastasis, recurrence and ERBB3 expression were associated with patient survival. The ERBB3 expression was not correlated to other clinicopathological factors. Furthermore, multivariate analysis revealed that ERBB3 expression was an independent prognostic factor influencing patient survival. Our results suggested that the expression of ERBB3 is associated with patient survival and could serve as a novel and valuable predictor for prognostic evaluation of patients with NPC.
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized