  • Search Results
  • Confocal imaging with a fluorescent bile acid analogue closely mimicking hepatic taurocholate disposition.
Confocal imaging with a fluorescent bile acid analogue closely mimicking hepatic taurocholate disposition.
Journal of pharmaceutical sciences (2014-03-22)
Tom De Bruyn, Wouter Sempels, Jan Snoeys, Nico Holmstock, Sagnik Chatterjee, Bruno Stieger, Patrick Augustijns, Johan Hofkens, Hideaki Mizuno, Pieter Annaert
ABSTRACT
This study aimed to characterize the in vitro hepatic transport mechanisms in primary rat and human hepatocytes of the fluorescent bile acid derivative N-(24-[7-(4-N,N-dimethylaminosulfonyl-2,1,3-benzoxadiazole)]amino-3α,7α,12α-trihydroxy-27-nor-5β-cholestan-26-oyl)-2'-aminoethanesulfonate (tauro-nor-THCA-24-DBD), previously synthesized to study the activity of the bile salt export pump (BSEP). The fluorescent bile acid derivative exhibited saturable uptake kinetics in suspended rat hepatocytes. Hepatic uptake was inhibited in the presence of substrates/inhibitors of the organic anion transporting polypeptide (Oatp) family and Na(+) -taurocholate cotransporting peptide (Ntcp). Concentration-dependent uptake of the fluorescent bile acid was also saturable in Chinese hamster ovary cells transfected with rNtcp, hNTCP, OATP1B1, or OATP1B3. The fluorescent bile acid derivative was actively excreted in the bile canaliculi of sandwich-cultured rat and human hepatocytes (SCRH and SCHH), with a biliary excretion index (BEI) of 26% and 32%, respectively. In SCRH, cyclosporin A significantly decreased the BEI to 5%. Quantification by real-time confocal imaging further confirmed canalicular transport of the fluorescent bile acid derivative (BEI = 75%). We conclude that tauro-nor-THCA-24-DBD is a useful probe to study interference of drugs with NTCP/Ntcp- and BSEP/Bsep-mediated transport in fluorescence-based in vitro assays.
Product Description
SAFC
Sodium butyrate
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine
Supelco
Sodium butyrate, certified reference material, TraceCERT®
Supelco
L-Glutamine, certified reference material, TraceCERT®
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)