  • Search Results
  • Characterization of the Dual Subcellular Localization of Lilium LsGRP1, a Plant Class II Glycine-Rich Protein.
Characterization of the Dual Subcellular Localization of Lilium LsGRP1, a Plant Class II Glycine-Rich Protein.
Phytopathology (2014-09-11)
Chia-Hua Lin, Chao-Ying Chen
ABSTRACT
The defense-related gene LsGRP1 exhibits an increased level of expression in Lilium spp. after being infected by Botrytis elliptica, the fungal pathogen of lily leaf blight. In this study, the expression profile of the LsGRP1 protein (a plant class II glycine-rich protein) was characterized biochemically and its subcellular localization in lily leaves was evaluated using immunohistochemistry, enhanced green fluorescent protein (EGFP) imaging, and protein extraction analysis. Using an LsGRP1-specific antibody, LsGRP1 was found to be most abundant in epidermal cells and phloem tissues. Leaves from lily plants at different growth stages demonstrated similar levels of 14- and 16-kDa LsGRP1 and a decreased amount of 23-kDa LsGRP1 at the senescence stage. LsGRP1-EGFP imaging and protein extraction assays revealed that 14-kDa LsGRP1 was located in the plasma membrane whereas 16- and 23-kDa LsGRP1 was weakly bound to the cell wall. The time course analyses of LsGRP1 expression in response to salicylic acid treatment or B. elliptica infection showed an increased accumulation of 14- and 23-kDa LsGRP1 over time. Because 23-kDa LsGRP1 could be detected by an ubiquitin antibody, conversion of 14-kDa to 23-kDa LsGRP1 via mono-ubiquitination was presumed, which is a phenomenon that has not been reported for a plant class II glycine-rich protein.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.