  • Search Results
  • Influence of excited-state absorption on time-resolved luminescence: general formalism and application to the phosphorescence of polycyclic aromatic hydrocarbons.
Influence of excited-state absorption on time-resolved luminescence: general formalism and application to the phosphorescence of polycyclic aromatic hydrocarbons.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-12-17)
Tiago Palmeira, Alexander Fedorov, Mário N Berberan-Santos
ABSTRACT
The luminescence decay of a species in an absorbing medium whose optical thickness changes with time, as occurs with triplet-triplet absorption following excitation cut-off, is studied theoretically and experimentally. A general luminescence decay function based on a distribution of optical thicknesses is presented. A simple decay function previously used empirically is shown to result from an exponential distribution of optical thicknesses. The general approach introduced allows the adequate description of the phosphorescence decays of two polycyclic aromatic hydrocarbons, coronene and triphenylene (normal and perdeuterated forms for both molecules), in polymer films in the presence of excited-state absorption.
Sigma-Aldrich
Coronene, purified by sublimation, 99%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Coronene, 97%
Coronene, BCR®, certified reference material
Triphenylene, BCR®, certified reference material
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Triphenylene, 98%
Supelco
Triphenylene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%